Skip to Content
Merck
All Photos(1)

Key Documents

574341

Sigma-Aldrich

Dimethyl sulfoxide-d6

99.9 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

(Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein:
1237248
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

0.42 mmHg ( 20 °C)

Quality Level

isotopic purity

99.9 atom % D

Assay

≥99.00%

form

liquid

autoignition temp.

573 °F

contains

0.03 % (v/v) TMS

expl. lim.

42 %

technique(s)

NMR: suitable

impurities

≤0.0250% water
water

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfoxide-d6 (DMSO-d6) is a deuterated NMR solvent containing 0.03% (v/v) TMS (tetramethylsilane). It undergoes photodissociation to generate CD3 radical photoproducts, which have been analyzed by infrared diode laserabsorption spectroscopy.[1] Dissociation dynamics of DMSO-d6 at 193nm was examined using photo fragment translational spectroscopy method.[2]

Application

Dimethyl sulfoxide-d6 may be used as an NMR solvent for 1H and 13C NMR experiments.[3]

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solution forms of an antitumor cyclic hexapeptide, RA-VII in dimethyl sulfoxide-d6 from nuclear magnetic resonance studies.
Itokawa H, et al.
Chemical & Pharmaceutical Bulletin, 40(4), 1050-1052 (1992)
Unraveling the dissociation of dimethyl sulfoxide following absorption at 193 nm.
Blank DA, et al.
J. Chem. Phys. , 106(2), 539-550 (1997)
Priya P Netalkar et al.
European journal of medicinal chemistry, 79, 47-56 (2014-04-12)
Air and moisture stable coordination compounds of late first row transition metals, viz. Co(II), Ni(II), Cu(II) and Zn(II), with a newly designed ligand, 2-(2-benzo[d]thiazol-2-yl)hydrazono)propan-1-ol (LH), were prepared and successfully characterized using various spectro-analytical techniques. The molecular structures of the ligand
Diode laser measurements of CD3 quantum yields and internal energy for the dissociation of dimethyl sulfoxide-d6.
Rudolph RN, et al.
J. Chem. Phys. , 106(4), 1346-1352 (1997)
Luís Novo et al.
Molecular pharmaceutics, 12(1), 150-161 (2014-11-11)
The applicability of small interfering RNA (siRNA) in future therapies depends on the availability of safe and efficient carrier systems. Ideally, siRNA delivery requires a system that is stable in the circulation but upon specific uptake into target cells can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service