Skip to Content
Merck
All Photos(2)

Documents

572403

Sigma-Aldrich

Cerium(IV) oxide-calcium doped

nanopowder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeO2
Molecular Weight:
172.11
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

form

nanopowder

contains

10 mol % calcium as dopant

composition

Ce0.9Ca0.1O2

reaction suitability

reagent type: catalyst
core: cerium

surface area

100-120 m2/g

particle size

<100 nm

SMILES string

[Ca].O=[Ce]=O

InChI

1S/Ca.Ce.2O.2H

InChI key

CBEFPOUMLUPIIH-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service