Skip to Content
Merck
All Photos(2)

Key Documents

569879

Sigma-Aldrich

3-(1-Thioureido)propyl, functionalized silica gel

200-400 mesh, extent of labeling: 1.1 mmol/g loading

Synonym(s):

Thiourea, silica gel supported

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$936.00

$936.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$936.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$936.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

reaction suitability

reagent type: catalyst
reagent type: chelator

extent of labeling

1.1 mmol/g loading

surface area

500 m2/g

particle size

200-400 mesh

pore size

60 Å pore size

SMILES string

CCCNC(N)=S

InChI

1S/C4H10N2S/c1-2-3-6-4(5)7/h2-3H2,1H3,(H3,5,6,7)

InChI key

UHGKYJXJYJWDAM-UHFFFAOYSA-N

Application

Metal Scavenger: Pd, Pt, Rh, Ru, and Hg

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service