Skip to Content
Merck
All Photos(1)

Key Documents

567833

Sigma-Aldrich

1-Isocyanato-5,6,7,8-tetrahydronaphthalene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO
CAS Number:
Molecular Weight:
173.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.5640 (lit.)

bp

264-265 °C (lit.)

density

1.112 g/mL at 25 °C (lit.)

SMILES string

O=C=Nc1cccc2CCCCc12

InChI

1S/C11H11NO/c13-8-12-11-7-3-5-9-4-1-2-6-10(9)11/h3,5,7H,1-2,4,6H2

InChI key

CGGOJVMAVIDVMK-UHFFFAOYSA-N

Caution

Refrigerate

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service