Skip to Content
Merck
All Photos(1)

Key Documents

566330

Sigma-Aldrich

9,9-Dihexylfluorene-2,7-diboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3B(OH)2C(CH2CH2CH2CH2CH2CH3)2C6H3B(OH)2
CAS Number:
Molecular Weight:
422.17
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%

form

solid

mp

>300 °C (lit.)

SMILES string

CCCCCCC1(CCCCCC)c2cc(ccc2-c3ccc(cc13)B(O)O)B(O)O

InChI

1S/C25H36B2O4/c1-3-5-7-9-15-25(16-10-8-6-4-2)23-17-19(26(28)29)11-13-21(23)22-14-12-20(27(30)31)18-24(22)25/h11-14,17-18,28-31H,3-10,15-16H2,1-2H3

InChI key

QFWOJNOZWMHNTK-UHFFFAOYSA-N

Application

Reactant for:
  • Rhodium-catalyzed hydroarylation
  • Molecular bridging of silicon nanogaps
  • Preparation of boronate ester coordinated polymer and macrocycle for sensing and storing of benzene
  • Synthesis of light-emitting carbazole-based copolymers bearing cyano-substituted arylenevinylene chromophore
  • Rhodium-catalyzed arylation

Other Notes

May contain varying amounts of anhydrides

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service