Skip to Content
Merck
All Photos(1)

Key Documents

566276

Sigma-Aldrich

Strontium bromide

anhydrous, beads, -10 mesh, 99.995%

Synonym(s):

Strontium dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrBr2
CAS Number:
Molecular Weight:
247.43
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.995%

form

beads

particle size

-10 mesh

SMILES string

Br[Sr]Br

InChI

1S/2BrH.Sr/h2*1H;/q;;+2/p-2

InChI key

YJPVTCSBVRMESK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Reagent for facile synthesis of poly(phenylcarbyne), a precursor to diamond-like carbon.

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Scherpelz et al.
Journal of chemical theory and computation, 12(8), 3523-3544 (2016-06-23)
We present an implementation of G0W0 calculations including spin-orbit coupling (SOC) enabling investigations of large systems, with thousands of electrons, and we discuss results for molecules, solids, and nanocrystals. Using a newly developed set of molecules with heavy elements (called

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service