Skip to Content
Merck
All Photos(1)

Key Documents

565636

Sigma-Aldrich

Sudan 408

Dye content ~95 %

Synonym(s):

1-(2-Methyl-4-o-tolylazo-phenylazo)-2-n-hexyloxy-naphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H32N4O
CAS Number:
Molecular Weight:
464.60
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, ~95%

mp

55.6-58.2 °C (lit.)

λmax

205 nm in ethanol

SMILES string

CCCCCCOc1ccc2ccccc2c1\N=N\c3ccc(cc3C)\N=N\c4ccccc4C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service