Skip to Content
Merck
All Photos(2)

Key Documents

565334

Sigma-Aldrich

2-Bromo-4-methylbenzaldehyde

technical grade

Synonym(s):

2-bromo-p-tolualdehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$33.60

$33.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$33.60

About This Item

Linear Formula:
Br(CH3)C6H3CHO
CAS Number:
Molecular Weight:
199.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$33.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

form

solid

mp

30-35 °C (lit.)

functional group

aldehyde
bromo

SMILES string

[H]C(=O)c1ccc(C)cc1Br

InChI

1S/C8H7BrO/c1-6-2-3-7(5-10)8(9)4-6/h2-5H,1H3

InChI key

MUZMDYCVUCMIDC-UHFFFAOYSA-N

Application

Reactant involved in:
  • Carbonylative Stille couplings followed by isomerization and intramolecular Heck reactions
  • Knoevenagel condensation
  • Cyclization of N-(α-methyl-p-methoxybenzyl)-imino-esters
  • Imination and oxidative heterocyclization / carbonylation
  • Intramolecular amination
  • Intermolecular and intramolecular hydroacylation of alkenes by aromatic aldehydes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service