Skip to Content
Merck
All Photos(2)

Key Documents

562521

Sigma-Aldrich

Cesium nitrate

Cabot high-purity grade

Synonym(s):

Nitric acid cesium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$351.00

$351.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Select a Size

Change View
1 KG
$351.00

About This Item

Empirical Formula (Hill Notation):
CsNO3
CAS Number:
Molecular Weight:
194.91
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

$351.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Quality Level

form

powder

mp

414 °C (lit.)

density

3.685 g/mL at 25 °C (lit.)

SMILES string

[Cs+].[O-]N(=O)=O

InChI

1S/Cs.NO3/c;2-1(3)4/q+1;-1

InChI key

NLSCHDZTHVNDCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of cesium nitratocuprate, Cs2[Cu(NO3)4], which crystallizes in a square-planar coordination around the Cu center.

Legal Information

Product of Cabot Performance Materials

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 1 - Repr. 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service