Skip to Content
Merck
All Photos(2)

Key Documents

56207

Sigma-Aldrich

1-Hydroxypiperidine

≥96.0% (GC)

Synonym(s):

1-Piperidinol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$178.00

$178.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$178.00

About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Beilstein:
102726
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$178.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Assay

≥96.0% (GC)

mp

37-39 °C

storage temp.

2-8°C

SMILES string

ON1CCCCC1

InChI

1S/C5H11NO/c7-6-4-2-1-3-5-6/h7H,1-5H2

InChI key

LKPFBGKZCCBZDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Base for Fmoc-based solid phase peptide synthesis

Light stabilizing agent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Y Wang et al.
Zhongguo yao li xue bao = Acta pharmacologica Sinica, 10(3), 252-256 (1989-05-01)
The biological N-oxidation of piperidine, a pharmacologically active biogenic amine of mammals and human beings, was studied in vitro. After incubation of piperidine-HCl in a fortified rat liver microsomal preparation (9000 x g supernatant) at 37 degrees C for 30
Akimitsu Miyaji et al.
Biochemical and biophysical research communications, 483(1), 178-182 (2017-01-04)
The dissociation of water molecules by ultrasound irradiation under aerobic conditions was demonstrated experimentally. To be able to detect the dissociation of water molecules, we performed the ultrasound irradiation of
Shipra Grover et al.
ACS infectious diseases, 7(1), 141-152 (2020-12-16)
MmpL3, an essential mycolate transporter in the inner membrane of Mycobacterium tuberculosis (Mtb), has been identified as a target of multiple, chemically diverse antitubercular drugs. However, several of these molecules seem to have secondary targets and inhibit bacterial growth by
Robert J Greenstein et al.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases, 13(5), e254-e263 (2009-03-24)
Without known mechanisms of action, thalidomide is used to treat a variety of non-malignant 'idiopathic' diseases. There is increasing concern that Mycobacterium avium subspecies paratuberculosis (MAP) may be zoonotic. Recently, methotrexate, azathioprine, 6-mercaptopurine (6-MP), 5-aminosalicylic acid (5-ASA), cyclosporine A, rapamycin
Areej Abuhammad et al.
PloS one, 7(12), e52790-e52790 (2013-01-04)
Latent M. tuberculosis infection presents one of the major obstacles in the global eradication of tuberculosis (TB). Cholesterol plays a critical role in the persistence of M. tuberculosis within the macrophage during latent infection. Catabolism of cholesterol contributes to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service