Skip to Content
Merck
All Photos(1)

Documents

561843

Sigma-Aldrich

N-Benzyl-N′-cyclohexylcarbodiimide, polymer-bound

100-200 mesh, extent of labeling: 1.0-2.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

N,N′-Dicyclohexylcarbodiimide, polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Quality Level

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: carboxyl reactive

extent of labeling

1.0-2.0 mmol/g loading

particle size

100-200 mesh

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Polymer-supported coupling reagent. Used in the preparation of a dicarboxamidocyclopentanol library.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weinshenker, G.
Organic Syntheses, 6, 951-951 (1988)
Synthesis of compound libraries based on 3,4-diaminocyclopentanol scaffolds.
Y Guan et al.
Journal of combinatorial chemistry, 2(4), 297-300 (2000-07-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service