Skip to Content
Merck
All Photos(1)

Documents

561037

Sigma-Aldrich

3,4-Difluorophenylmagnesium bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3MgBr
CAS Number:
Molecular Weight:
217.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

bp

65 °C

density

0.965 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Fc1ccc([Mg]Br)cc1F

InChI

1S/C6H3F2.BrH.Mg/c7-5-3-1-2-4-6(5)8;;/h1,3-4H;1H;/q;;+1/p-1

InChI key

WZHAVYGMKGPNFN-UHFFFAOYSA-M

Application

3,4-Difluorophenylmagnesium bromide can be used:
  • To prepare 3,4-difluorophenylboronic acid, a key building block employed in the synthesis of norepinephrine transporter selective molecules.
  • In the synthesis of trimethoxybenzophenone derived tubulin polymerization inhibitors.
  • In one of the key synthetic steps for the preparation of diarylketoxime derivatives as potent MCH-1R antagonists.
  • In the synthesis of biologically important hydroxybupropion analogs useful in smoking cessation.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of in vitro and in vivo profiles of hydroxybupropion analogues: aids to smoking cessation
Lukas RJ, et al.
Journal of Medicinal Chemistry, 53(12), 4731-4748 (2010)
Synthesis and monoamine transporter binding properties of 3α-(substituted phenyl) nortropane-2β-carboxylic acid methyl esters. Norepinephrine transporter selective compounds
Carroll FI, et al.
Journal of medicinal chemistry, 48(11), 3852-3857 (2005)
Discovery of novel diarylketoxime derivatives as selective and orally active melanin-concentrating hormone 1 receptor antagonists
Suzuki T, et al.
Bioorganic & medicinal chemistry letters, 19(18), 5339-5345 (2009)
2?Amino?3, 4, 5?Trimethoxybenzophenones as Potent Tubulin Polymerization Inhibitors.
Chuang H Y, et al.
ChemMedChem, 6(3), 450-456 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service