Recommended Products
Assay
97%
refractive index
n20/D 1.3550 (lit.)
bp
29-30 °C (lit.)
storage temp.
2-8°C
SMILES string
FC(F)(F)C(Br)=C
InChI
1S/C3H2BrF3/c1-2(4)3(5,6)7/h1H2
InChI key
QKBKGNDTLQFSEU-UHFFFAOYSA-N
General description
2-Bromo-3,3,3-trifluoro-1-propene can be added to N2 gas in order to enhance its fire suppression efficiency.
Application
2-Bromo-3,3,3-trifluoro-1-propene may be used in the synthesis of 3,3,3 -trifluoropropynyllithium anion via treatment with lithium diisopropylamide at a very low temperature(-78oC).
accessory
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2 - STOT SE 3
Target Organs
Central nervous system, Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nucleophilic addition of 3, 3, 3-trifluoropropynyllithium to d-glyceraldimine: Concise synthesis of both enantiomers of 5, 5, 5-trifluoronorvaline
Journal of Fluorine Chemistry, 128(10), 1182-1186 (2007)
Synthesis of trifluoromethylated and gem-difluoromethylenated biologically interesting compounds from fluorine-containing synthons
Synlett, 2011(08), 1052-1072 (2011)
Fire protection with bromoalkene/nitrogen gaseous mixtures
Industrial & Engineering Chemistry Research, 40(21), 4649-4653 (2001)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service