Skip to Content
Merck
All Photos(6)

Key Documents

560871

Sigma-Aldrich

2-Methyl-2-propanesulfinamide

97%, racemic

Synonym(s):

(±)-tert-Butylsulfinamide, 1,1-Dimethylethylsulfinamide, 2-Methyl-2-propanesulfinamide, 2-Methylpropan-2-sulfinamide, tert-Butanesulfinamide, tert-Butylsulfinamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$158.00
25 G
$585.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$158.00
25 G
$585.00

About This Item

Linear Formula:
(CH3)3CS(O)NH2
CAS Number:
Molecular Weight:
121.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

97-101 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)S(N)=O

InChI

1S/C4H11NOS/c1-4(2,3)7(5)6/h5H2,1-3H3

InChI key

CESUXLKAADQNTB-UHFFFAOYSA-N

Application

Synthesis of 4-sulfonyliminopiperidine and its reaction with benzyl Grignard to form an important pharmaceutical building block, 4-benzyl-4-aminopiperidne.[1] Also used to prepare spiro tosylaziridines via methylene insertion into the derived sulfonylimine.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synlett, 833-833 (2006)
Synlett, 2565-2565 (2006)

Articles

Ellman's sulfinamide is available in both enantiomeric and racemic forms for your research. This versatile and useful auxiliary has found extensive use both in academics and industry.

Related Content

Ellman group developed electron-rich phosphine ligands for C-H functionalization and tert-Butanesulfinamide for asymmetric amine synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service