Skip to Content
Merck
All Photos(1)

Key Documents

560170

Sigma-Aldrich

2-Bromo-4-methylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH3)C6H3COOH
CAS Number:
Molecular Weight:
215.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

143-147 °C (lit.)

SMILES string

Cc1ccc(C(O)=O)c(Br)c1

InChI

1S/C8H7BrO2/c1-5-2-3-6(8(10)11)7(9)4-5/h2-4H,1H3,(H,10,11)

InChI key

ZZYYOHPHSYCHQG-UHFFFAOYSA-N

General description

2-Bromo-4-methylbenzoic acid can be prepared from 2-bromo-4-methylbenzonitrile via hydrolysis.[1]

Application

2-Bromo-4-methylbenzoic acid may be used to synthesize:
  • 4′-hydroxy-5:6′-dimethyldibenzo-α-pyrone[2]
  • 2-bromo-4-methylbenzophenone[1]
  • 7-hydroxy-5′-methyl-6-n-hexyl-3:4-benzocoumarin[3]
  • 2-(trimethylsilyl)ethyl 2-bromo-4-methylbenzoate[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies in the biochemistry of micro-organisms. 90. Alternariol and alternariol monomethyl ether, metabolic products of Alternaria tenuis.
Raistrick H, et al.
The Biochemical Journal, 55(3), 421-421 (1953)
204. Tetrahydrodibenzopyran derivatives isomeric with tetrahydrocannabinols.
Avison AWD, et al.
Journal of the Chemical Society, 952-955 (1949)
A Radioiodinated MIBG-octreotate conjugate exhibiting enhanced uptake and retention in sstr2-expressing tumor cells.
Vaidyanathan G, et al.
Bioconjugate Chemistry, 18(6), 2122-2130 (2007)
Intramolecular Reactions. IV. Chain Transfer Reaction Involving an Aromatic Hydrogen Atom and Related Reactions of 2-(4'-Methylbenzoyl)-benzenediazonium Salts1.
DeTar DF and Relyea DI.
Journal of the American Chemical Society, 78(17), 4302-4305 (1955)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service