Skip to Content
Merck
All Photos(1)

Key Documents

559512

Sigma-Aldrich

2′-Chloro-5′-(trifluoromethyl)acetophenone

97%

Synonym(s):

2-Chloro-5-(trifluoromethyl)acetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CF3)COCH3
CAS Number:
Molecular Weight:
222.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Assay

97%

refractive index

n20/D 1.4810 (lit.)

bp

204-205 °C (lit.)

density

1.392 g/mL at 25 °C (lit.)

functional group

chloro
fluoro
ketone

SMILES string

CC(=O)c1cc(ccc1Cl)C(F)(F)F

InChI

1S/C9H6ClF3O/c1-5(14)7-4-6(9(11,12)13)2-3-8(7)10/h2-4H,1H3

InChI key

YRGBMTWHOFQSDJ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

196.0 °F - closed cup

Flash Point(C)

91.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service