Skip to Content
Merck
All Photos(2)

Key Documents

558893

Sigma-Aldrich

1-Ethyl-4-ethynylbenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C6H4C≡CH
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.5420 (lit.)

bp

30 °C/0.05 mmHg (lit.)

density

0.930 g/mL at 25 °C (lit.)

SMILES string

CCc1ccc(cc1)C#C

InChI

1S/C10H10/c1-3-9-5-7-10(4-2)8-6-9/h1,5-8H,4H2,2H3

InChI key

ZNTJVJSUNSUMPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-4-ethynylbenzene undergoes homocoupling reaction in the presence of copper(I) chloride (catalyst) and air (oxidant) to yield symmetrical 1,4-disubstituted 1,3-diyne.[1]

Application

1-Ethyl-4-ethynylbenzene may be used to synthesize:[2]
  • 2,4-bis[(4-ethylphenyl)ethynyl]-6-methoxy-1,3,5-triazine
  • 2-[(4-ethylphenyl)ethynyl]-4-methoxy-6-(phenylethynyl)-1,3,5-triazine
  • 2-chloro-4-[(4-ethylphenyl)ethynyl]-6-methoxy-1,3,5-triazine
  • 4-chloro-6-[(4-ethylphenyl)ethynyl]-N,N-diphenyl-1,3,5-triazin-2-amine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

160.0 °F - closed cup

Flash Point(C)

71.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CuCl-catalyzed green oxidative alkyne homocoupling without palladium, ligands and bases.
Yin K, et al.
Green Chemistry, 13(3), 591-593 (2011)
Linear and V-Shaped Alkynyl-1, 3, 5-triazines.
Gao P, et al.
European Journal of Organic Chemistry, 32, 7020-7027 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service