Skip to Content
Merck
All Photos(1)

Key Documents

557986

Sigma-Aldrich

(R)-(−)-2-Decanol

97%, optical purity97% (ee) (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$182.00

$182.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$182.00

About This Item

Linear Formula:
CH3(CH2)7CH(OH)CH3
CAS Number:
Molecular Weight:
158.28
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$182.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

optical activity

[α]20/D −8.0°, c = 1% in chloroform

optical purity

97% (ee) (HPLC)

refractive index

n20/D 1.4340 (lit.)

bp

211 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCCC[C@@H](C)O

InChI

1S/C10H22O/c1-3-4-5-6-7-8-9-10(2)11/h10-11H,3-9H2,1-2H3/t10-/m1/s1

InChI key

ACUZDYFTRHEKOS-SNVBAGLBSA-N

Application

(R)-(−)-2-Decanol can be used as a chiral inductor in photoelectrocyclization of tropolone methyl ether.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced stereoselectivity in photoelectrocyclization of tropolone ethers via confinement in chiral inductor-modified lyotropic liquid crystals
Lv F, et al.
Organic Letters, 10(16), 3473-3476 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service