Skip to Content
Merck
All Photos(1)

Key Documents

557358

Sigma-Aldrich

2-Methyl-1,5-hexadiene

97%

Synonym(s):

2-Methyldiallyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2CH2C(CH3)=CH2
CAS Number:
Molecular Weight:
96.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

97%

refractive index

n20/D 1.417 (lit.)
n20/D 1.4170 (lit.)

bp

92 °C (lit.)

density

0.712 g/mL at 25 °C (lit.)

functional group

allyl

SMILES string

CC(=C)CCC=C

InChI

1S/C7H12/c1-4-5-6-7(2)3/h4H,1-2,5-6H2,3H3

InChI key

SLQMKNPIYMOEGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-1,5-hexadiene on mercury-photosensitized irradiation, undergoes internal cycloaddition to afford 1-methylbicyclo[2.1.1]hexane (as major product).[1] High-temperature pyrolysis of 2-methyl-1,5-hexadiene affords 1,5-hexadiene and 2,5-dimethyl-1,5-hexadiene.[2] 2-Methyl-1,5-hexadiene undergoes stereoselective and regioselective reaction with dialkylaluminum chloride (Et2AlCl) and titanium alkoxide [Ti(OPr-i)4, a catalyst] followed by oxidation to afford the corresponding five-membered 3-alkylcycloalkyl methanol.[3] 2-Methyl-1,5-hexadiene is formed as a major product during the condensation reaction between allyl chloride and methallyl chloride in the presence of magnesium.[4]

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Flam. Liq. 2 - Muta. 1B - STOT RE 2

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vapor-phase thermolyses of 3-hydroxy-1, 5-hexadienes. II. Effects of methyl substitution.
Negishi EI, et al.
Journal of the American Chemical Society, 116(18), 8404-8405 (1994)
Olefins and Diolefins from Allylic Chlorides.
Henne AL, et al.
Journal of the American Chemical Society, 63(12), 3474-3476 (1941)
Mercury (3P1) photosensitized internal cycloaddition reactions in 1, 4-, 1, 5-, and 1, 6-dienes.
Srinivasan R and Carlough KH.
Journal of the American Chemical Society, 89(19), 4932-4936 (1967)
Vapor-phase thermolyses of 3-hydroxy-1, 5-hexadienes. II. Effects of methyl substitution.
Viola A, et al.
Journal of the American Chemical Society, 89(14), 3462-3470 (1967)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service