Skip to Content
Merck
All Photos(3)

Key Documents

557315

Sigma-Aldrich

4-(Methylsulfonyl)benzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2C6H4SO2Cl
CAS Number:
Molecular Weight:
254.71
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

165-169 °C (lit.)

SMILES string

CS(=O)(=O)c1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C7H7ClO4S2/c1-13(9,10)6-2-4-7(5-3-6)14(8,11)12/h2-5H,1H3

InChI key

TYJOQICPGZGYDT-UHFFFAOYSA-N

Application

4-(Methylsulfonyl)benzenesulfonyl chloride may be used in the preparation of 5′-deoxy-5′-[4-(methylsulfonyl)benzenesulfonamido]thymidine.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of inhibitors of thymidine monophosphate kinase from Bacillus anthracis.
Byun Y, et al.
Nucleosides, nucleotides & nucleic acids, 27(3), 244-260 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service