Skip to Content
Merck
All Photos(2)

Key Documents

556602

Sigma-Aldrich

4-Bromobenzenesulfonic acid monohydrate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$187.00

$187.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$187.00

About This Item

Linear Formula:
BrC6H4SO3H · H2O
CAS Number:
Molecular Weight:
255.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$187.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

90-93 °C (lit.)

functional group

bromo
sulfonic acid

SMILES string

O.OS(=O)(=O)c1ccc(Br)cc1

InChI

1S/C6H5BrO3S.H2O/c7-5-1-3-6(4-2-5)11(8,9)10;/h1-4H,(H,8,9,10);1H2

InChI key

JHULXEZFSDUIRI-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service