Skip to Content
Merck
All Photos(1)

Key Documents

555843

Sigma-Aldrich

3,5-Diacetyl-2,4,6-trimethyl-1,4-dihydropyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO2
CAS Number:
Molecular Weight:
207.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

154-159 °C (lit.)

SMILES string

CC1C(C(C)=O)=C(C)NC(C)=C1C(C)=O

InChI

1S/C12H17NO2/c1-6-11(9(4)14)7(2)13-8(3)12(6)10(5)15/h6,13H,1-5H3

InChI key

BFHJJFUZJSTSBY-UHFFFAOYSA-N

General description

3,5-Diacetyl-2,4,6-trimethyl-1,4-dihydropyridine [3,5-diacetyl-1,4-dihydroxy-2,4,6-trimethylpyridine] can be obtained by reacting acetyl acetone, corresponding aldehyde, NH3 gas.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Photochemistry of Novel 3, 5-Diacetyl-1, 4-dihydropyridines. II [1].
Memarian HR, et al.
Monatshefte fur Chemie / Chemical Monthly, 135(7), 833-838 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service