Skip to Content
Merck
All Photos(1)

Key Documents

555347

Sigma-Aldrich

Cap Mix A (90% Tetrahydrofuran, 10% Acetic anhydride)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

impurities

≤0.01% (water)

Quality Level

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service