Skip to Content
Merck
All Photos(1)

Key Documents

553867

Sigma-Aldrich

2,4-Dichloro-5-fluoro-3-nitrobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6HCl2FNO2COOH
CAS Number:
Molecular Weight:
254.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

193-197 °C (lit.)

functional group

carboxylic acid
chloro
fluoro
nitro

SMILES string

OC(=O)c1cc(F)c(Cl)c(c1Cl)[N+]([O-])=O

InChI

1S/C7H2Cl2FNO4/c8-4-2(7(12)13)1-3(10)5(9)6(4)11(14)15/h1H,(H,12,13)

InChI key

PCSAPCNEJUEIGU-UHFFFAOYSA-N

General description

2,4-Dichloro-5-fluoro-3-nitrobenzoic acid can be prepared from 2,4-dichloro-5-fluorobenzoic acid.[1]

Application

2,4-Dichloro-5-fluoro-3-nitrobenzoic acid may be used to synthesize:
  • ethyl 3-(N,N-dimethylamino)-2-(2,4-dichloro-5-fluoro-3-nitrobenzoyl)acrylate[2]
  • 7-chloro-1-cyclopropyl-6-fluoro-8-nitro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid[3][4]
  • ethyl-3-(N,N-dimethylamino)-2-(2,4-dichloro-5-fluoro-3-nitrobenzoyl) acrylate[5]
  • substituted 4(1H)-oxoquinoline-3-carboxylic acid[6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Engler et al.
Antimicrobial agents and chemotherapy, 42(5), 1151-1159 (1998-05-21)
Photodegradation of sparfloxacin was observed by means of high-pressure liquid chromatography with UV detection and liquid chromatography coupled with UV detection and tandem mass spectrometry (LC-MS/MS). Three products were detected. Comparison with an independently synthesized derivative of sparfloxacin revealed the
Yusuf M Al-Hiari et al.
Molecules (Basel, Switzerland), 13(11), 2880-2893 (2008-11-21)
[1,4]Diazepino[2,3-h]quinolone carboxylic acid 3 and its benzo-homolog tetrahydroquino[7,8-b]benzodiazepine-3-carboxylic acid 5 were prepared via PPAcatalyzed thermal lactamization of the respective 8-amino-7-substituted-1,4-dihydroquinoline-3-carboxylic acid derivatives 8, 10. The latter compounds were obtained by reduction of their 8-nitro-7-substituted-1,4-dihydroquinoline-3-carboxylic acid precursors 7, 9 which, in
Mohammed H Al-Huniti et al.
Molecules (Basel, Switzerland), 12(8), 1558-1568 (2007-10-26)
Substituted [1,4]thiazepino[2,3-h]quinolinecarboxylic acid 3 is prepared by PPA-catalyzed thermal lactamization of the respective 8-amino-7-[(2-carboxyethyl)thio]-1,4-dihydroquinoline-3-carboxylic acid 9. The latter synthon is obtained by reduction of the 8-nitro-1,4-dihydroquinoline precursor 8 which, in turn, is made accessible via interaction of 3-mercaptopropionic acid with
Synthesis and antibacterial activity of 9-cyclopropyl-4-fluoro-6-oxo-6, 9-dihydro-[1, 2, 5] thiadiazolo [3, 4-h] quinoline-7-carboxylic acid and its ethyl ester.
Al-Qawasmeh RA, et al.
ARKIVOC (Gainesville, FL, United States), 12, 322-326 (2009)
Synthesis and antibacterial properties of new 8-nitrofluoroquinolone derivatives.
Al-Hiari YM, et al.
Molecules (Basel), 12(6), 1240-1258 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service