Skip to Content
Merck
All Photos(1)

Key Documents

55262

Sigma-Aldrich

4-[4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy]butyric acid

≥98.0% (HPLC), for peptide synthesis

Synonym(s):

Hydroxyethyl photolinker

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,100.00

$1,100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$1,100.00

About This Item

Empirical Formula (Hill Notation):
C13H17NO7
CAS Number:
Molecular Weight:
299.28
Beilstein:
7717635
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$1,100.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

4-[4-(1-Hydroxyethyl)-2-methoxy-5-nitrophenoxy]butyric acid, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

form

powder

reaction suitability

reagent type: cross-linking reagent

mp

163-166 °C

SMILES string

COc1cc(C(C)O)c(cc1OCCCC(O)=O)[N+]([O-])=O

InChI

1S/C13H17NO7/c1-8(15)9-6-11(20-2)12(7-10(9)14(18)19)21-5-3-4-13(16)17/h6-8,15H,3-5H2,1-2H3,(H,16,17)

InChI key

DUIJUTBRRZCWRD-UHFFFAOYSA-N

Other Notes

Photolabile linker which can be bound to amino- and hydroxymethyl resins. Synthesis of sensitive acids[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher P. Holmes
The Journal of organic chemistry, 62(8), 2370-2380 (1997-04-18)
Both a model phenacyl and o-nitrobenzyl photolabile linker from the literature along with four new o-nitrobenzyl linkers were prepared and the kinetics of their photolytic cleavage examined in solution. The linkers were prepared by amidation of the carboxylic acid anchoring
D.L. Whitehouse et a
Tetrahedron Letters, 38, 7851-7851 (1997)
Mime Kobayashi et al.
Journal of biomaterials science. Polymer edition, 30(13), 1161-1171 (2019-06-16)
We have developed biocompatible scaffolds that enable cell fate control with visible light. The scaffolds are based on synthetic collagen-like polypeptide, poly(prolyl-hydroxyprolyl-glycyl) {poly(Pro-Hyp-Gly)} which has been used for cosmetics and other healthcare applications. Bioactive peptides were conjugated to the scaffolds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service