Skip to Content
Merck
All Photos(1)

Key Documents

552534

Sigma-Aldrich

(S)-(−)-2-(Diphenylmethyl)pyrrolidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N
CAS Number:
Molecular Weight:
237.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

solid

optical activity

[α]24/D −3.0°, c = 1% in chloroform

refractive index

n20/D 1.587 (lit.)
n20/D 1.5870 (lit.)

bp

135 °C/0.01 mmHg (lit.)

density

1.062 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CN[C@@H](C1)C(c2ccccc2)c3ccccc3

InChI

1S/C17H19N/c1-3-8-14(9-4-1)17(16-12-7-13-18-16)15-10-5-2-6-11-15/h1-6,8-11,16-18H,7,12-13H2/t16-/m0/s1

InChI key

OXOBKZZXZVFOBB-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used as excellent chiral solvating agents to determine the enantiomeric composition of chiral carboxylic acids directly by NMR analysis.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bailey, D. J. et al.
Tetrahedron Asymmetry, 8, 149-149 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service