Skip to Content
Merck
All Photos(1)

Key Documents

550760

Sigma-Aldrich

3-Iodobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4CHO
CAS Number:
Molecular Weight:
232.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

57-60 °C (lit.)

functional group

aldehyde
iodo

storage temp.

2-8°C

SMILES string

Ic1cccc(C=O)c1

InChI

1S/C7H5IO/c8-7-3-1-2-6(4-7)5-9/h1-5H

InChI key

RZODAQZAFOBFLS-UHFFFAOYSA-N

General description

3-Iodobenzaldehyde can be prepared by the iodination of benzaldehyde using 1,3-diiodo-5,5-dimethylhydantoin in sulfuric acid.[1]

Application

3-Iodobenzaldehyde may be used as a starting material in the preparation of:
  • 3-iodocinnamic acid[2]
  • 3-(3-hydroxy-3-methylbut-1-yn-1-yl)benzaldehyde[3]
  • 1,3-dihydroxy-2-(3-iodophenyl)-4,4,5,5-tetramethylimidazolidine[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzuki Reactions with Stable Organic Radicals-Synthesis of Biphenyls Substituted with Nitronyl-Nitroxide Radicals.
Stroh C, et al.
European Journal of Organic Chemistry, 2005(17), 3697-3703 (2005)
Synthesis and evaluation of potent and selective human V1a receptor antagonists as potential ligands for PET or SPECT imaging.
Fabio K, et al.
Bioorganic & Medicinal Chemistry, 20(3), 1337-1345 (2012)
Cross-coupling of aryl iodides with paramagnetic terminal acetylenes derived from 4, 4, 5, 5-tetramethyl-2-imidazoline-1-oxyl 3-oxide.
Klyatskaya SV, et al.
Russian Chemical Bulletin, 51(1), 128-134 (2002)
1, 3-Diiodo-5,5-dimethylhydantoin-An efficient reagent for iodination of aromatic compounds.
Chaikovskii VK, et al.
Russ. J. Org. Chem., 43(9), 1291-1296 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service