Skip to Content
Merck
All Photos(1)

Key Documents

550647

Sigma-Aldrich

2-Nitro-4-thiocyanatoaniline

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(SCN)NH2
CAS Number:
Molecular Weight:
195.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

113 °C (lit.)

SMILES string

Nc1ccc(SC#N)cc1[N+]([O-])=O

InChI

1S/C7H5N3O2S/c8-4-13-5-1-2-6(9)7(3-5)10(11)12/h1-3H,9H2

InChI key

QUWHIBBGKKRYFW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of 2-Nitro-4-thiocyanatoaniline with 4-Nitrochlorobenzene.
Pilyugin VS, et al.
Russ. J. Gen. Chem., 72(9), 1439-1443 (2002)
Synthesis of 2-Amino-5 (6)-(4-aminophenyl) benzimidazole Derivatives: II. Reaction of 2-Nitro-4-thiocyanatoaniline with 4-Nitrochlorobenzene.
Pilyugin VS, et al.
Russ. J. Org. Chem., 39(7), 979-984 (2003)
Andreas Schmidt et al.
Organic & biomolecular chemistry, 1(23), 4342-4350 (2003-12-20)
2,3-Diaminophenol 4, 3,4-diaminophenol 5, 4-methoxy-1,2-diaminobenzene 6, 3,4-diaminobenzenethiol 7, 2,3-diaminobenzoic acid 8, and 3,4-diaminobenzoic acid 9 were reacted with 2,4-pentanedione to yield the corresponding benzo[b][1,4]diazepinium salts, respectively. The hydroxy-benzo[b][1,4]diazepinium salts 17 and 18 do not form mesomeric betaines (MB) on deprotonation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service