Skip to Content
Merck
All Photos(3)

Key Documents

549975

Sigma-Aldrich

3-(Diethylamino)propylamine

≥99%

Synonym(s):

N,N-Diethyl-1,3-diaminopropane, N,N-Diethyl-1,3-propanediamine, DEAPA

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$41.40
500 ML
$99.70
2 L
$298.00

$41.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$41.40
500 ML
$99.70
2 L
$298.00

About This Item

Linear Formula:
(C2H5)2N(CH2)3NH2
CAS Number:
Molecular Weight:
130.23
Beilstein:
741879
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$41.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

4.4 (vs air)

vapor pressure

19.5 mmHg ( 70 °C)

Assay

≥99%

autoignition temp.

419 °F

expl. lim.

11.6 %

refractive index

n20/D 1.4416 (lit.)

bp

159 °C (lit.)

density

0.826 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCN(CC)CCCN

InChI

1S/C7H18N2/c1-3-9(4-2)7-5-6-8/h3-8H2,1-2H3

InChI key

QOHMWDJIBGVPIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Diethylamino)propylamine (3-(diethylamino)-1-propylamine, DEAPA) is a diamine.

Application

3-(Diethylamino)propylamine may be used in the preparation of:
  • Biocompatible polyampholyte-coated (poly(acrylic acid)-co-3-(diethylamino)-propylamine magnetite nanoparticles (PAMNPs).[1]
  • Hyaluronic acid grafted with 3-diethylaminopropyl (DEAP) group (HA-g-DEAP) nanoparticles for use as a drug delivery vehicle.[2]
  • DEAPA-modified poly(vinyl alcohol) (PVA) polymers that can be employed in gene silencing.[3]
It may also be used to introduce cationic groups during the surface modification of microfibrillated cellulose (MFC).[4]

Legal Information

Product of Arkema Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical surface modifications of microfibrillated cellulose.
Stenstad P, et al.
Cellulose, 15(1), 35-45 (2008)
Juliane Nguyen et al.
Pharmaceutical research, 27(12), 2670-2682 (2010-09-18)
The objective of this study was to investigate how the degree of amine substitution of amine-modified poly(vinyl alcohol) (PVA) affects complexation of siRNA, protection of siRNA against degrading enzymes, intracellular uptake and gene silencing. A series of DEAPA-PVA polymers with
Low toxicity and long circulation time of polyampholyte-coated magnetic nanoparticles for blood pool contrast agents.
Wang Qi, et al.
Scientific Reports, 5, 7774-7774 (2015)
Hyaluronated nanoparticles with pH-and enzyme-responsive drug release properties.
Kim SW, et al.
Colloids and Surfaces. B, Biointerfaces, 116, 359-364 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service