Skip to Content
Merck
All Photos(1)

Documents

547085

Sigma-Aldrich

Z-trans-4-Hydroxy-L-prolinol

Synonym(s):

Benzyl (2S,4R)-4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO4
CAS Number:
Molecular Weight:
251.28
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

optical activity

[α]20/D −33°, c = 15 in chloroform

Quality Level

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.547 (lit.)

bp

240 °C (lit.)

density

1.297 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC[C@@H]1C[C@@H](O)CN1C(=O)OCc2ccccc2

InChI

1S/C13H17NO4/c15-8-11-6-12(16)7-14(11)13(17)18-9-10-4-2-1-3-5-10/h1-5,11-12,15-16H,6-9H2/t11-,12+/m0/s1

InChI key

WDEQGLDWZMIMJM-NWDGAFQWSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service