Skip to Content
Merck
All Photos(2)

Key Documents

545554

Sigma-Aldrich

6-Acetylthiohexanoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COS(CH2)5CO2H
CAS Number:
Molecular Weight:
190.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.491 (lit.)

density

1.144 g/mL at 25 °C (lit.)

functional group

carboxylic acid
thioester

SMILES string

CC(=O)SCCCCCC(O)=O

InChI

1S/C8H14O3S/c1-7(9)12-6-4-2-3-5-8(10)11/h2-6H2,1H3,(H,10,11)

InChI key

PIFIISOIFYMVLS-UHFFFAOYSA-N

General description

6-Acetylthiohexanoic acid (6-ATH), also known as 6-(acetylsulfanyl)hexanoic acid, is a hexanoic acid derivative that contains thioacetate group.

Application

6-Acetylthiohexanoic acid has been used in a study to estimate its minimal inhibitory concentration (MIC) by microdilution assay. It may be used as a precursor for generating poly-3-hydroxy-6-acetylthiohexanoateco-4-acetylthiobutanoate (PHACOS).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (3-hydroxyalkanoate) s: diversification and biomedical applications: a state of the art review.
Hazer DB, et al.
Materials Science & Engineering. C, Materials For Biological Applications, 32(4), 637-647 (2012)
Nina Dinjaski et al.
Biomaterials, 35(1), 14-24 (2013-10-08)
Biomaterial-associated infections represent a significant clinical problem, and treatment of these microbial infections is becoming troublesome due to the increasing number of antibiotic-resistant strains. Here, we report a naturally functionalized bacterial polyhydroxyalkanoate (PHACOS) with antibacterial properties. We demonstrate that PHACOS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service