Skip to Content
Merck
All Photos(1)

Key Documents

544604

Sigma-Aldrich

2,5-Di(tert-butylperoxy)-2,5-dimethyl-3-hexyne

technical grade, 90%

Synonym(s):

2,5-Bis(tert-butylperoxy)-2,5-dimethyl-3-hexyne, 2,5-Dimethyl-2,5-di(tert-butylperoxy)-2-hexyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COOC(CH3)2CC(CH3)2OOC(CH3)3
CAS Number:
Molecular Weight:
286.41
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical grade

Quality Level

Assay

90%

composition

active oxygen content, 10 wt. %

t1/2

10 hr(125 °C)

density

0.883 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OOC(C)(C)C#CC(C)(C)OOC(C)(C)C

InChI

1S/C16H30O4/c1-13(2,3)17-19-15(7,8)11-12-16(9,10)20-18-14(4,5)6/h1-10H3

InChI key

ODBCKCWTWALFKM-UHFFFAOYSA-N

Application

Free radical polymerization initiator.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Org. Perox. B - Skin Irrit. 2

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service