Skip to Content
Merck
All Photos(1)

Key Documents

544574

Sigma-Aldrich

Cobalt naphthenate

6%

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$89.40
1 KG
$271.00

$89.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$89.40
1 KG
$271.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$89.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

concentration

≤6.5% Co+2 in mineral spirits
6%

refractive index

n20/D 1.4620

density

0.921 g/mL at 25 °C

SMILES string

[Co+2].[O-]C(=O)CCC2CC(CC2)CC.[O-]C(=O)CCC1CC(CC1)CC

InChI

1S/2C10H18O2.Co/c2*1-2-8-3-4-9(7-8)5-6-10(11)12;/h2*8-9H,2-7H2,1H3,(H,11,12);/q;;+2/p-2

InChI key

XSWKLHINRKWMTD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cobalt naphthenate is the catalyst that promotes the curing process and allows effective cross-linking for a variety of polymeric films.[1]

Application

Cobalt naphthenate may be used as a catalyst that facilitates the free radical polymerization of (3-methacryloxypropyl)trimethoxysilane (MPS), which can be used to functionalize alumina nanoparticles.[2] It can be used as a promoter for the development of unsaturated flame retardant polyesters.[3]

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 3 - Repr. 2 - Skin Sens. 1 - STOT RE 1 - STOT RE 1 Oral

Target Organs

Central nervous system, Gastrointestinal tract

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.0 °F - Tag closed cup

Flash Point(C)

48.9 °C - Tag closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of artificial urushi via ring-opening reaction of benzoxazine with renewable cardanol
Ma H, et al.
Polymer, 132, 41-50 (2017)
Highly thermostable and durably flame-retardant unsaturated polyester modified by a novel polymeric flame retardant containing Schiff base and spirocyclic structures
Zhao D, et al.
Chemical Engineering Journal, 344(27), 419-430 (2018)
Surface functionalized alumina nanoparticle filled polymeric nanocomposites with enhanced mechanical properties
Guo Z, et al.
Journal of Materials Chemistry, 16(27), 2800-2808 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service