Skip to Content
Merck
All Photos(1)

Key Documents

544531

Sigma-Aldrich

2-Bromo-5-nitrothiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2BrNO2S
CAS Number:
Molecular Weight:
208.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

44-48 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Br)s1

InChI

1S/C4H2BrNO2S/c5-3-1-2-4(9-3)6(7)8/h1-2H

InChI key

ZPNFMDYBAQDFDY-UHFFFAOYSA-N

General description

2-Bromo-5-nitrothiophene is a heteroaryl halide that can be prepared from thiophene by bromination followed by nitration.[1] It participates in the synthesis of oligothiophene precursors for generating (porphinato)zinc(II)-based chromophores.[2]

Application

2-Bromo-5-nitrothiophene may be used in the preparation of:
  • 3-bromo-4-(5-nitro-2-thienyl)-6-ethoxypyridine[3]
  • 5-chloro-2-methoxy-4-(5-nitrothien-2-yl)-pyridine[4]
  • 5-(5-nitrothien-2-yl)pyrimidine[5]
  • 2-methoxy-5-(5-nitrothien-2-yl)pyrimidine[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oligo (2,5?thienyleneethynylene) s with Terminal Donor?Acceptor Substitution.
Meier H, et al.
European Journal of Organic Chemistry, 2006(2), 405-413 (2006)
New Shelf-stable halo-and alkoxy-substituted Pyridylboronic acids and their Suzuki cross-coupling reactions to yield heteroarylpyridines.
Parry PR, et al.
Synthesis, 2003(07), 1035-1038 (2003)
5-Pyrimidylboronic acid and 2-methoxy-5-pyrimidylboronic acid: new heteroarylpyrimidine derivatives via Suzuki cross-coupling reactions.
Saygili N, et al.
Organic & Biomolecular Chemistry, 2(6), 852-857 (2004)
Design, synthesis, linear, and nonlinear optical properties of conjugated (porphinato) zinc (II)-based donor-acceptor chromophores featuring nitrothiophenyl and nitrooligothiophenyl electron-accepting moieties.
Zhang TG, et al.
Journal of the American Chemical Society, 127(27), 9710-9720 (2005)
Functionalized pyridylboronic acids and their Suzuki cross-coupling reactions to yield novel heteroarylpyridines
Parry PR, et al.
The Journal of Organic Chemistry, 67(21), 7541-7543 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service