Skip to Content
Merck
All Photos(1)

Key Documents

540420

Sigma-Aldrich

[(1E)-1-Methoxy-2-nitrovinyl]methylamine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NCH=C(OCH3)NHCH3
CAS Number:
Molecular Weight:
132.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

116-120 °C (lit.)

storage temp.

2-8°C

SMILES string

CN\C(OC)=C/[N+]([O-])=O

InChI

1S/C4H8N2O3/c1-5-4(9-2)3-6(7)8/h3,5H,1-2H3/b4-3+

InChI key

ISROKTUWQRVBQO-ONEGZZNKSA-N

Caution

Refrigerate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service