Skip to Content
Merck
All Photos(1)

Documents

53695

Sigma-Aldrich

3-(Tritylthio)propionic acid

≥97.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3CSCH2CH2COOH
CAS Number:
Molecular Weight:
348.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (T)

SMILES string

OC(=O)CCSC(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C22H20O2S/c23-21(24)16-17-25-22(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20/h1-15H,16-17H2,(H,23,24)

InChI key

AECGEIVNZGQBJT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naglaa Salem El-Sayed et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-14)
Cell-penetrating peptide [WR]₅ has been previously shown to be an efficient molecular transporter for various hydrophilic and hydrophobic molecules. The peptide was synthesized using Fmoc/tBu solid-phase chemistry, and one arginine was replaced with one lysine to enable the conjugation with
Mathew R Schnorenberg et al.
ACS omega, 3(10), 14144-14150 (2018-11-10)
Despite the therapeutic promise of phospholipid-based nanocarriers, a major obstacle to their widespread clinical translation is a susceptibility to fatty acid ester hydrolysis, leading to lack of quality control and inconsistencies in self-assembly formulations. Using electrospray ionization mass spectrometry fragmentation
Scott H Medina et al.
Journal of controlled release : official journal of the Controlled Release Society, 209, 317-326 (2015-05-17)
Many lead molecules identified in drug discovery campaigns are eliminated from consideration due to poor solubility and low cell permeability. These orphaned molecules could have clinical value if solubilized and delivered properly. SVS-1 is a de novo designed peptide that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service