Skip to Content
Merck
All Photos(1)

Key Documents

536903

Sigma-Aldrich

1,2,2,6,6-Pentamethyl-4-piperidone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19NO
CAS Number:
Molecular Weight:
169.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.476 (lit.)

bp

230-231 °C (lit.)

density

0.949 g/mL at 25 °C (lit.)

functional group

ketone

storage temp.

−20°C

SMILES string

CN1C(C)(C)CC(=O)CC1(C)C

InChI

1S/C10H19NO/c1-9(2)6-8(12)7-10(3,4)11(9)5/h6-7H2,1-5H3

InChI key

GHJUORCGZFHNKG-UHFFFAOYSA-N

General description

1,2,2,6,6-Pentamethyl-4-piperidone is an intermediate for preparing pentamethylpiperidines of pharmacological importance.[1] Its conformational analysis based on proton magnetic resonance spectra, dipole moments and molar Kerr constant have been reported.[2]

Application

1,2,2,6,6-Pentamethyl-4-piperidone may be used in the preparation of 1-phenyl-2,2,6,6-tetramethyl-4-phenyliminopiperidine and 1-benzyl-2,2,6,6-tetramethyl-4-benzyliminopiperidine.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular polarisability. The molecular conformations of some substituted 4-piperidones as solutes.
Aroney MJ, et al.
Journal of the Chemical Society B: Physical Organic, 98-102 (1966)
Synthesis and pharmacological investigation of 2,2,6,6-tetramethyl-4-aryl(Aralkyl) aminopiperidines.
Nikit-skaya ES, et al.
Pharmaceutical Chemistry Journal, 8(2), 82-86 (1974)
Rubiralta M, et al.
Piperidine: Structure, Preparation, Reactivity, and Synthetic Applications of Piperidine and its Derivatives, 394-394 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service