534226
4′-(2-Chloroacetyl)acetanilide
97%
Synonym(s):
4-(2-Chloroacetyl)acetanilide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ClCH2COC6H4NHCOCH3
CAS Number:
Molecular Weight:
211.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
97%
mp
216-219 °C (lit.)
SMILES string
CC(=O)Nc1ccc(cc1)C(=O)CCl
InChI
1S/C10H10ClNO2/c1-7(13)12-9-4-2-8(3-5-9)10(14)6-11/h2-5H,6H2,1H3,(H,12,13)
InChI key
VMMDOCYDNRLESP-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service