Skip to Content
Merck
All Photos(1)

Key Documents

533262

Sigma-Aldrich

2-Fluoro-6-methylpyridine

97%

Synonym(s):

6-Fluoro-2-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6FN
CAS Number:
Molecular Weight:
111.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.47 (lit.)

bp

140-141 °C (lit.)

density

1.077 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(F)n1

InChI

1S/C6H6FN/c1-5-3-2-4-6(7)8-5/h2-4H,1H3

InChI key

UDMNVTJFUISBFD-UHFFFAOYSA-N

General description

2-Fluoro-6-methylpyridine, also known as 2-fluoro-6-picoline, can be prepared via diazotization of 2-amino-6-methylpyridine in hydrogen fluoride containing 40% pyridine solution. [1] The effect of substituents on spectral properties of 2-fluoro-6-methylpyridine has been investigated based on 13C NMR, UV and IR spectral data.[2]

Application

2-Fluoro-6-methylpyridine may be used in the preparation of:
  • 2-fluoro-6-pyridinecarboxylic acid[3]
  • 2,2′-[1-(6-methylpyridin-2-yl)ethane-1,1-diyl]dipyridine[4]
  • 2-fluoropyridine-6-aldoxime[5]
  • 2-fluoro-6-(dibromomethyl)pyridine[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.1 °F - closed cup

Flash Point(C)

66.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of Geometric Isomers of a Dicopper Complex by Using a 19F-Labeled Ligand: Dynamics, Structures, and DFT Calculations.
Durot S, et al.
Inorganic Chemistry, 49(17), 7832-7840 (2010)
Synthesis of some fluorine-containing pyridinealdoximes of potential use for the treatment of organophosphorus nerve-agent poisoning.
Timperley CM, et al.
Journal of Fluorine Chemistry, 132(8), 541-547 (2011)
Efficient synthesis of pentakis-and tris (pyridine) ligands.
Unal EA, et al.
Tetrahedron Letters, 53(1), 54-55 (2012)
A facile preparation of fluoropyridines from aminopyridines via diazotization and fluorodediazoniation in HF or HF-pyridine solutions.
Fukuhara T, et al.
Journal of Fluorine Chemistry, 38(3), 435-438 (1988)
The synthesis of 2-fluoro-4-and 2-fluoro-6-pyridinecarboxylic acid and derivatives.
Roe A, et al.
Journal of the American Chemical Society, 71(12), 4152-4153 (1949)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service