Skip to Content
Merck
All Photos(2)

Key Documents

532312

Sigma-Aldrich

p-Toluenesulfonic acid, polymer-bound

Macroporous, 30-60 mesh, extent of labeling: 2.0-3.0 mmol/g loading

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$413.00
100 G
$435.00

$413.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$413.00
100 G
$435.00

About This Item

MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$413.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

2.0-3.0 mmol/g loading

particle size

30-60 mesh

SMILES string

C=Cc1ccccc1.OS(=O)(=O)c2ccccc2.C=Cc3ccc(C=C)cc3

InChI key

LGTBNVJSBQBEJG-UHFFFAOYSA-N

Application

Scavenges nitrogen nucleophiles, activated ester electrophiles, and bases such as EDC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chen J. et al.
Tetrahedron Letters, 40, 9195-9195 (1999)
Flynn, D.L.
Journal of the American Chemical Society, 119, 4874-4874 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service