Skip to Content
Merck
All Photos(1)

Documents

532207

Sigma-Aldrich

2-Chloro-N-methylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NHCH3
CAS Number:
Molecular Weight:
141.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.579 (lit.)

bp

218 °C (lit.)

density

1.15 g/mL at 25 °C (lit.)

SMILES string

CNc1ccccc1Cl

InChI

1S/C7H8ClN/c1-9-7-5-3-2-4-6(7)8/h2-5,9H,1H3

InChI key

WGNNILPYHCKCFF-UHFFFAOYSA-N

General description

2-Chloro-N-methylaniline (2-Cl-N-MA), also known as N-methyl-2-chloroaniline, is a secondary amine. The iridium-catalyzed borylation of 2-Cl-N-MA affords the correponding ortho-borylated derivative.

Application

2-Chloro-N-methylaniline (N-Methyl-2-chloroaniline) may be used in the synthesis of 2-chloro-N-methylaniline-blocked polyisocyanate and N-methyl-N-nitro-2-chloroaniline.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amine?blocked polyisocyanates. I. Synthesis of novel N?methylaniline?blocked polyisocyanates and deblocking studies using hot?stage fourier transform infrared spectroscopy.
Sankar G and Nasar AS.
Journal of Polymer Science Part A: Polymer Chemistry, 45(8), 1557-1570 (2007)
Molecular and crystal structures of N-methyl-N-nitro-2-chloroaniline.
Prezhdo VV, et al.
Crystallography Reports, 46(3), 389-393 (2001)
Silyl-directed, iridium-catalyzed ortho-borylation of arenes. A one-pot ortho-borylation of phenols, arylamines, and alkylarenes.
Boebel TA and Hartwig JF.
Journal of the American Chemical Society, 130(24), 7534-7535 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service