Skip to Content
Merck
All Photos(1)

Key Documents

531979

Sigma-Aldrich

(S,S)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminoaluminum chloride

Synonym(s):

(S,S)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminoaluminum(III) chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$251.00

$251.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$251.00

About This Item

Empirical Formula (Hill Notation):
C36H52AlClN2O2
CAS Number:
Molecular Weight:
607.24
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$251.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

mp

>350 °C (lit.)

Quality Level

SMILES string

CC(C)(C)c1cc2\C=N\[C@H]3CCCC[C@@H]3\N=C\c4cc(cc(c4O[Al](Cl)Oc2c(c1)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C36H54N2O2.Al.ClH/c1-33(2,3)25-17-23(31(39)27(19-25)35(7,8)9)21-37-29-15-13-14-16-30(29)38-22-24-18-26(34(4,5)6)20-28(32(24)40)36(10,11)12;;/h17-22,29-30,39-40H,13-16H2,1-12H3;;1H/q;+3;/p-3/b37-21+,38-22+;;/t29-,30-;;/m0../s1

InChI key

KGVBCKDZFMLRMA-RUIQGICGSA-K

Application

(S,S)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminoaluminum chloride can be used as a chiral catalyst:
  • In the preparation of substituted hydroxyalkyl aminooxazole derivatives by reacting isocyanides with aldehydes.[1]
  • In the cyanation of α,β-unsaturated imides to yield saturated imides.[2][3]

Legal Information

Sold under license from Sterling Pharma Solutions Limited.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Glenn M Sammis et al.
Journal of the American Chemical Society, 126(32), 9928-9929 (2004-08-12)
Cooperative heterobimetallic catalysis was used as a design principle to achieve a highly reactive system for the enantioselective conjugate addition of cyanide to alpha,beta-unsaturated imides. A dual-catalyst pathway involving chiral (salen)Al complex 1b and chiral (pybox)Er complex 4b provides measurable
Highly enantioselective, catalytic conjugate addition of cyanide to alpha, beta-unsaturated imides
Sammis GM and Jacobsen EN
Journal of the American Chemical Society, 125(15), 4442-4443 (2003)
Shi-Xin Wang et al.
Organic letters, 9(18), 3615-3618 (2007-08-10)
In the presence of a catalytic amount of (salen)Al(III)Cl complex (4e), reaction between alpha-isocyanoacetamides (1) and aldehydes (2) afforded the corresponding 5-aminooxazoles (3) in good yields and enantioselectivity.

Articles

The first reports of salen used as ligands with manganese for asymmetric epoxidation reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service