Skip to Content
Merck
All Photos(3)

Key Documents

53166

Sigma-Aldrich

3-(Triphenylphosphonio)propane-1-sulfonate

≥99.0% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$689.00

$689.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$689.00

About This Item

Empirical Formula (Hill Notation):
C21H21O3PS
CAS Number:
Molecular Weight:
384.43
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$689.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

≥99.0% (HPLC)

impurities

≤1.0% water
≤100 mg/kg halogene (as chloride)

functional group

phosphine
sulfonic acid

SMILES string

[O-]S(=O)(=O)CCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H21O3PS/c22-26(23,24)18-10-17-25(19-11-4-1-5-12-19,20-13-6-2-7-14-20)21-15-8-3-9-16-21/h1-9,11-16H,10,17-18H2

InChI key

CHVFGCXPLGAAQN-UHFFFAOYSA-N

General description

3-(Triphenylphosphonio)propane-1-sulfonate is a zwitterionic, sulfonic acid-functionalized ionic liquid.[1]

Application

3-(Triphenylphosphonio)propane-1-sulfonate has been used as a zwitterion to study its effect on the work function of indium tin oxide (ITO) in inverted polymer solar cells (PSCs).[2] It can be used in the preparation of triphenyl-(3-sulfopropyl)phosphonium phosphotungstate, an efficient green catalyst for esterification reactions.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient, inverted polymer solar cells with indium tin oxide modified with solution-processed zwitterions as the transparent cathode.
Sun K, et al.
ACS Applied Materials & Interfaces, 4(4), 2009-2017 (2012)
Task-Specific Ionic Liquids
Aldrich Chemfiles, 5(6), 17-17 (2006)
Phosphotungstic acid salt of triphenyl (3-sulfopropyl) phosphonium: An efficient and reusable solid catalyst for esterification.
Zhang W, et al.
Catalysis Communications, 11(3), 151-154 (2009)

Articles

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service