Skip to Content
Merck
All Photos(1)

Key Documents

531464

Sigma-Aldrich

2-Iodophenylacetonitrile

97%

Synonym(s):

2-Iodobenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4CH2CN
CAS Number:
Molecular Weight:
243.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.618 (lit.)

bp

113-120 °C/0.5 mmHg (lit.)

density

1.75 g/mL at 25 °C (lit.)

functional group

iodo
nitrile

SMILES string

Ic1ccccc1CC#N

InChI

1S/C8H6IN/c9-8-4-2-1-3-7(8)5-6-10/h1-4H,5H2

InChI key

FPSGTRJUQLYLHE-UHFFFAOYSA-N

General description

2-Iodophenylacetonitrile is a 2-aryl substituted nitrile. It reacts with lactams to form ring-fused isoquinolinones via palladium-catalyzed carboxamidation in tandem with aldol condensation.[1]

Application

2-Iodophenylacetonitrile may be used in the preparation of:
  • 2?-aminobiphen-2-ylacetonitrile[2]
  • ethyl (2-iodophenyl)iminoacetate hydrochloride[3]
  • 3,4-disubstituted 2-naphthalenamines[4]

It may also be used in the preparation of the following nitriles:[5]
  • 2-(2-iodophenyl)-2-methylpropanenitrile
  • 1-(2-iodophenyl)cyclopentanecarbonitrile
  • 5-bromo-2-(2-iodophenyl)pentanenitrile
  • 2-(2-iodophenyl)-2-propylpentanenitrile
  • 1-(2-iodophenyl)cyclohexanecarbonitrile
  • 1-(2-Iodophenyl)cyclopropanecarbonitrile

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Crich et al.
The Journal of organic chemistry, 71(9), 3452-3463 (2006-04-22)
The [1-cyano-2-(2-iodophenyl)]ethylidene group is introduced as an acetal-protecting group for carbohydrate thioglycoside donors. The group is easily introduced under mild conditions, over short reaction times, and in the presence of a wide variety of other protecting groups by the reaction
Studies in Acyl C? H Activation via Aryl and Alkyl to Acyl ?Through Space? Migration of Palladium.
Kesharwani T, et al.
Organic Letters, 11(12), 2591-2593 (2009)
Palladium-catalyzed carboxamidation reaction and aldol condensation reaction cascade: A facile approach to ring-fused isoquinolinones.
Chouhan G and Alper H.
Organic Letters, 10(21), 4987-4990 (2008)
Hirokazu Tsukamoto et al.
The Journal of organic chemistry, 81(5), 1733-1745 (2015-11-26)
1,2-Bis(diphenylphosphino)ethane (dppe)-ligated palladium(II) complexes catalyze the annulation of internal alkynes with 2-(cyanomethyl)phenylboronates to provide 3,4-disubstituted-2-naphthalenamines in good yields. The annulation reaction proceeds under mild and neutral conditions and requires methanol as an essential solvent. In addition to symmetrical alkynes, unsymmetrical
Palladium-catalyzed borylation of ortho-substituted phenyl halides and application to the one-pot synthesis of 2,2'-disubstituted biphenyls.
Baudoin O, et al.
The Journal of Organic Chemistry, 65(26), 9268-9271 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service