Skip to Content
Merck
All Photos(1)

Documents

530581

Sigma-Aldrich

1-Vinylnaphthalene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH=CH2
CAS Number:
Molecular Weight:
154.21
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

contains

3000 ppm tert-butylcatechol as inhibitor

impurities

≤2% poly(1-vinylnaphthalene)

refractive index

n20/D 1.653 (lit.)

bp

135-138 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

C=Cc1cccc2ccccc12

InChI

1S/C12H10/c1-2-10-7-5-8-11-6-3-4-9-12(10)11/h2-9H,1H2

InChI key

IGGDKDTUCAWDAN-UHFFFAOYSA-N

Application

1-Vinylnaphthalene may be used in the synthesis of 1-naphthyl-1,2-ethanediol, which can further be utilized as a metal hydrogenated catalyst. It may also be used as a substrate material to produce azidocyanation based products.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F

Flash Point(C)

101 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modulation of Trehalose Dimycolate and Immune System by Rv0774c Protein Enhanced the Intracellular Survival of
Arbind Kumar et al.
Frontiers in cellular and infection microbiology, 7, 289-289 (2017-07-18)
Rana Dhar et al.
Immunopharmacology and immunotoxicology, 40(1), 43-51 (2017-12-05)
Immune dysregulation has been implicated in the pathogenesis of many diseases. Macrophages play a crucial role contributing to the onset, progression, and resolution of inflammation. Macrophage inflammatory mediators are of considerable interest as potential targets to treat inflammatory diseases. The
1-Naphthyl-1, 2-ethanediol as a new chiral modifier of platinum in the enantioselective hydrogenation of activated ketones.
Marinas A, et al.
J. Catal., 221(2), 666-669 (2004)
Enantioselective Copper-Catalyzed Intermolecular Amino-and Azidocyanation of Alkenes in a Radical Process.
Wang D, et al.
Angewandte Chemie (International Edition in English), 56(8), 2054-2058 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service