Skip to Content
Merck
All Photos(1)

Key Documents

528943

Sigma-Aldrich

4-Fluoro-2-methoxyphenol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$144.00

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$144.00

About This Item

Linear Formula:
FC6H3(OCH3)OH
CAS Number:
Molecular Weight:
142.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.517 (lit.)

bp

195 °C (lit.)

density

1.247 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

COc1cc(F)ccc1O

InChI

1S/C7H7FO2/c1-10-7-4-5(8)2-3-6(7)9/h2-4,9H,1H3

InChI key

OULGLTLTWBZBLO-UHFFFAOYSA-N

General description

4-Fluoro-2-methoxyphenol is a fluorinated methoxy-substituted catechol analog.[1]

Application

4-Fluoro-2-methoxyphenol may be used in the synthesis of:
  • 4-halo-masked o-benzoquinones (MOBs)[2]
  • fluorinated masked o-benzoquinone[3]
  • poly(4-fluoro-2-methoxyphenol)[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

214.9 °F - closed cup

Flash Point(C)

101.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diels-Alder reactions of halogenated masked o-benzoquinones: synthesis of halogen-substituted bicyclo [2.2. 2] octenones.
Tetrahedron Letters, 50(7), 773-775 (2009)
Ana Carolina de Almeida et al.
European journal of pharmacology, 660(2-3), 445-453 (2011-04-19)
Apocynin, a methoxy-substituted catechol (4-hydroxy-3-methoxyacetophenone), originally extracted from the roots of Picrorhiza kurroa, has been extensively used as a non-toxic inhibitor of the multienzymatic complex NADPH oxidase. We discovered that the analogous methoxy-substituted catechol, 4-Fluoro-2-methoxyphenol (F-apocynin), in which the acetyl
Novel photoconductive polyfluorophenol synthesized by an enzyme.
Zaragoza-Gasca P, et al.
Journal of Molecular Catalysis. B, Enzymatic, 72(1), 25-27 (2011)
A fluorinated masked o-benzoquinone.
Patrick TB, et al.
Journal of Fluorine Chemistry, 125(12), 1965-1966 (2007)
P K Chakraborty et al.
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes, 42(7), 673-681 (1991-01-01)
The synthesis of 4-[18F]fluoroguaiacol (4-[18F]fluoro-2-methoxyphenol) has been achieved in no-carrier-added form starting from 2-methoxy-4-nitrobenzaldehyde, using nucleophilic aromatic substitution by [18F]fluoride followed by Baeyer-Villiger oxidation of the benzaldehyde to the phenol. Demethylation with boron tribromide gave 4-[18F]fluorocatechol (1,2-dihydroxy-4-[18F]fluorobenzene) with an overall

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service