Skip to Content
Merck
All Photos(1)

Key Documents

52792

Sigma-Aldrich

(2R,5R)-2,5-Hexanediol

≥99.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O2
CAS Number:
Molecular Weight:
118.17
Beilstein:
1719251
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D −35±2°, c = 9% in chloroform

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

mp

52-54 °C

storage temp.

2-8°C

SMILES string

C[C@@H](O)CC[C@@H](C)O

InChI

1S/C6H14O2/c1-5(7)3-4-6(2)8/h5-8H,3-4H2,1-2H3/t5-,6-/m1/s1

InChI key

OHMBHFSEKCCCBW-PHDIDXHHSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chiral building for the synthesis of C2 symmetric chiral ligands, e.g. (S,S)-2,5-dimethylpyrrolidine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Nagai et al.
Synlett, 289-289 (1994)
A Practical Synthesis of (2R,5R)-2,5-Hexanediol.
P. Saravanan et al.
The Journal of organic chemistry, 62(8), 2669-2670 (1997-04-18)
M.J. Kim et al.
Synlett, 767-767 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service