Skip to Content
Merck
All Photos(1)

Key Documents

527823

Sigma-Aldrich

2,4,5-Trimethylbenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2CN
CAS Number:
Molecular Weight:
145.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

57-61 °C (lit.)

SMILES string

Cc1cc(C)c(cc1C)C#N

InChI

1S/C10H11N/c1-7-4-9(3)10(6-11)5-8(7)2/h4-5H,1-3H3

InChI key

VTCFOBGVVUVYHP-UHFFFAOYSA-N

Application

2,4,5-Trimethylbenzonitrile may be used as a probe to locate the Co2+ and dimers [Co-O-Co]2+ in the Co-Na-MOR and Co-H-MOR (MOR = a zeolite].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Location of Isolated Co2+ and [Co- O- Co]2+ in Co-MOR as Investigated by Means of FTIR with Acetonitrile and 2, 4, 5-Trimethylbenzonitrile as Probe Molecules.
Pietrogiacomi D, et al.
The Journal of Physical Chemistry C, 114(41), 17812-17818 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service