Skip to Content
Merck
All Photos(1)

Key Documents

524670

Sigma-Aldrich

Luperox® DHD-9, 2-Butanone peroxide solution

~32 wt. % in phthalate-free plasticizer mixture

Synonym(s):

2-Butanone peroxide solution, Methyl ethyl ketone peroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(OOH)(CH3)OOC(OOH)(CH3)C2H5
CAS Number:
Molecular Weight:
210.22
Beilstein:
1759757
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

reaction suitability

reagent type: oxidant

concentration

~32 wt. % in phthalate-free plasticizer mixture

refractive index

n20/D 1.437

density

0.99 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCC(C)(OO)OOC(C)(CC)OO

InChI

1S/C8H18O6/c1-5-7(3,11-9)13-14-8(4,6-2)12-10/h9-10H,5-6H2,1-4H3

InChI key

WFUGQJXVXHBTEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polymerization initiator

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Org. Perox. D - Repr. 2 - Skin Corr. 1B

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 1

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allergic contact dermatitis due to methyl ethyl ketone peroxide, cobalt naphthenate and acrylates in the manufacture of fibreglass-reinforced plastics.
Keiko Minamoto et al.
Contact dermatitis, 46(1), 58-59 (2002-03-29)
Yan-Fu Lin et al.
Journal of hazardous materials, 153(3), 1071-1077 (2007-11-09)
Runaway reactions by methyl ethyl ketone peroxide (MEKPO) are an important issue in Asia, due to its unstable structure and extensive heat release during upset situations. This study employed differential scanning calorimetry (DSC) to draw the experimental data for MEKPO
M V S Subbalaxmi et al.
The National medical journal of India, 23(3), 150-151 (2010-10-19)
Methyl ethyl ketone peroxide, commonly used in the lamination industry, is a highly inflammable substance. Ingestion of methyl ethyl ketone peroxide is associated with a high morbidity and mortality. It can cause injury to the gastrointestinal tract on ingestion, leading
N Bates et al.
Pediatrics, 108(2), 473-476 (2001-08-03)
A 6-year-old boy developed respiratory distress, metabolic acidosis, severe esophageal and gastric burns, and a coagulopathy after ingestion of an unknown volume of methyl ethyl ketone peroxide (MEKP) in dimethyl phthalate. He was discharged from the pediatric intensive care unit
[Chemical burns of the upper digestive tract and cytolytic hepatitis following ingestion of a mixture of methyl ethyl ketone peroxide, diacetone alcohol and diisobutyl phthalate].
D Chataigner et al.
Journal de toxicologie clinique et experimentale, 12(4-5), 205-206 (1992-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service