Skip to Content
Merck
All Photos(2)

Key Documents

523380

Sigma-Aldrich

4-[(Trimethylsilyl)ethynyl]benzaldehyde

97%

Synonym(s):

4-(Trimethylsilylethynyl)benzaldehyde, 4-[2-(Trimethylsilyl)ethynyl]benzaldehyde, p-[(Trimethylsilyl)ethynyl]benzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$319.00

$319.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$319.00

About This Item

Linear Formula:
(CH3)3SiC≡CC6H4CHO
CAS Number:
Molecular Weight:
202.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$319.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

66-70 °C (lit.)

functional group

aldehyde

SMILES string

C[Si](C)(C)C#Cc1ccc(C=O)cc1

InChI

1S/C12H14OSi/c1-14(2,3)9-8-11-4-6-12(10-13)7-5-11/h4-7,10H,1-3H3

InChI key

UZQDUXAJFTWMDT-UHFFFAOYSA-N

General description

4-[(Trimethylsilyl)ethynyl]benzaldehyde can be synthesized by reacting 4-bromobenzaldehyde and triphenylphosphine in anhydrous triethylamine with ethynyltrimethylsilane and then with palladium(II) acetate under argon.[1] It can also be prepared by Sonogashira coupling reaction between 4-bromobenzaldehyde and (trimethylsilyl) ethynyl.[2]

Application

4-[(Trimethylsilyl)ethynyl]benzaldehyde may be used in the preparation of the following:
  • 4,4,5,5-tetramethyl-2-{4-[(trimethylsilyl)ethynyl]phenyl}imidazolidine-1,3-diol[3]
  • 5,10,15-triphenyl-20-{4-[2-(trimethylsilyl)ethynyl]phenyl}porphyrin[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the potential of porphyrin-spiked triarylamine stars for bulk heterojunction solar cells.
Kengthanomma T, et al.
Journal of Material Chemistry A, 1(35), 10524-10531 (2013)
Facile synthesis of ethynylated benzoic acid derivatives and aromatic compounds via ethynyltrimethylsilane.
Austin WB, et al.
The Journal of Organic Chemistry, 46(11), 2280-2286 (1981)
Linear porphyrin dimers with fluorenyl arms linked by an ethynyl bridge.
Merhi A, et al.
Tetrahedron, 69(34), 7112-7124 (2013)
Synthesis of covalently bonded nanostructure from two porphyrin molecular wires leading to a molecular tube.
Ishida T, et al.
Tetrahedron Letters, 47(30), 5265-5268 (2006)
Cascade Reactions of Me3Si-Substituted Imidazolidine-1, 3-Diols with PbO2, Including Oxidation of the Corresponding Diol and Subsequent Elimination of the Trimethylsilyl Fragment.
Tretyakov E, et al.
European Journal of Organic Chemistry, 2007(22), 3639-3647 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service